EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Anionic Oligothiophenes Compete for Binding of X-34 but not PIB to Recombinant Aβ Amyloid Fibrils and Alzheimer's Disease Brain-Derived Aβ.
PMID 27767229
Abstract
Deposits comprised of amyloid-β (Aβ) are one of the pathological hallmarks of Alzheimer's disease (AD) and small hydrophobic ligands targeting these aggregated species are used clinically for the diagnosis of AD. Herein, we observed that anionic oligothiophenes efficiently displaced X-34, a Congo Red analogue, but not Pittsburgh compound B (PIB) from recombinant Aβ amyloid fibrils and Alzheimer's disease brain-derived Aβ. Overall, we foresee that the oligothiophene scaffold offers the possibility to develop novel high-affinity ligands for Aβ pathology only found in human AD brain, targeting a different site than PIB.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1954 X-34, ≥90% (HPLC)
C24H18O6