EMAIL THIS PAGE TO A FRIEND
Journal of chemical information and modeling
A Multiscale Study on the Penetration Enhancement Mechanism of Menthol to Osthole.
PMID 27768312
Abstract
Menthol is a widely used penetration enhancer in clinical medicine due to its high efficiency and relative safety. However, details of the penetration enhancement mechanism of menthol on the molecular level is rarely involved in the discussion. In this work, the penetration enhancement (PE) mechanism of menthol is explored by a multiscale method containing molecular dynamics simulations, in vitro penetration experiments, and transmission electron microscopy. Osthole is chosen to be the tested drug due to its common use in external preparations and because it often accompanies menthol as a PE in the preparations. The results show that menthol in each testing concentration can impair the lipid packing of stratum corneum (SC) and promote osthole permeating into SC, and the penetration promoting effect has an optimal concentration. At a low concentration, menthol causes the bilayer to relax with a reduction in thickness and increment in the lipid headgroup area. At a high concentration, menthol destroys the bilayer structure of SC and causes lipids to form a reversed micelle structure. The penetration enhancement mechanism of menthol is characterized mainly by the disruption of the highly ordered SC lipid in low concentrations and an improvement in the partitioning of drugs into the SC in high concentrations. The results can provide some assistance for additional studies and applications of menthol as a penetration enhancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31185
D.E.R. 332, used as embedding medium
C21H24O4