EMAIL THIS PAGE TO A FRIEND
Journal of the National Cancer Institute
Evolution in vitro of stemlines with minimal karyotypic deviations in a human heteroploid cell line.
PMID 277716
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87090201
C32
96020925
C32TG