EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
N-Glycoform-dependent interactions of megalin with its ligands.
PMID 27773703
Abstract
Megalin is a 600-kDa single-spanning transmembrane glycoprotein and functions as an endocytic receptor, distributed not only in the kidney but also in other tissues. Structurally and functionally distinct ligands for megalin have been identified. Megalin has 30 potential N-glycosylation sites in its extracellular domain. We found that megalin interacts with its ligands in a glycoform-dependent manner. Distribution of megalin and glycans was histochemically analyzed in mouse kidneys. Kidney absorption of Cy5-labeled ligands was examined in vivo. Megalin-ligand interactions were analyzed using ligand blotting and ELISA. Megalins expressed on renal proximal convoluted tubules (PCTs) and proximal straight tubules (PSTs) have different N-glycans. PCT megalin stained with Lens culinaris agglutinin (LCA), which recognizes core-fucosyl N-glycans catalyzed by α1,6-fucosyltransferase (Fut8). In contrast, PST megalin stained with wheat germ agglutinin (WGA), which recognizes hybrid-type N-glycans. Retinol-binding protein-Cy5 (RBP-Cy5) was endocytosed by megalin on PCTs but minimally endocytosed by PSTs. BSA-Cy5 was endocytosed nearly equally by both tubules. The purified LCA-positive glycoform megalin had higher binding activity for RBP and vitamin D-binding protein than did WGA-positive glycoform megalin. Both glycoforms had nearly the same BSA- and kanamycin-binding activities. RBP-binding analysis of megalin lacking core fucose, in Fut8(-/-) mouse kidneys, had significantly decreased binding activity. N-Glycosylation of megalin can modulate its ligand-binding activity. Core fucosylation, in particular, is a modification crucial for megalin-RBP interactions. Cell type-specific glycoforms of megalin exist in the proximal tubular cells and modulate ligand absorption capacity.