EMAIL THIS PAGE TO A FRIEND
Leukemia
The LIN28B/let-7 axis is a novel therapeutic pathway in multiple myeloma.
PMID 27773931
Abstract
MYC is a major oncogenic driver of multiple myeloma (MM) and yet almost no therapeutic agents exist that target MYC in MM. Here we report that the let-7 biogenesis inhibitor LIN28B correlates with MYC expression in MM and is associated with adverse outcome. We also demonstrate that the LIN28B/let-7 axis modulates the expression of MYC, itself a let-7 target. Further, perturbation of the axis regulates the proliferation of MM cells in vivo in a xenograft tumor model. RNA-sequencing and gene set enrichment analyses of CRISPR-engineered cells further suggest that the LIN28/let-7 axis regulates MYC and cell cycle pathways in MM. We provide proof of principle for therapeutic regulation of MYC through let-7 with an LNA-GapmeR (locked nucleic acid-GapmeR) containing a let-7b mimic in vivo, demonstrating that high levels of let-7 expression repress tumor growth by regulating MYC expression. These findings reveal a novel mechanism of therapeutic targeting of MYC through the LIN28B/let-7 axis in MM that may impact other MYC-dependent cancers as well.