EMAIL THIS PAGE TO A FRIEND
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie
Pseudo-accommodation in non-amblyopic children after bilateral cataract surgery and implantation with a monofocal intraocular lens: prevalence and possible mechanisms.
PMID 27785598
Abstract
Some pseudophakic patients implanted with a monofocal intraocular lens (IOL) have good near visual acuity (VA) with their distance correction. The objective was to evaluate the prevalence of pseudo-accommodation in children after bilateral cataract surgery, without amblyopia, and to define its mechanisms. Observational study that took place in a pediatric ophthalmology department, Paris, France. A total of 68 eyes were included, 40 from 23 children and 28 from 14 adults, with a corrected distance VA above 20/25 and a normal near VA (20/25) with +3 addition. Pseudo-accommodation was defined as a near VA better than 20/50 with the distance correction and without addition. Prevalence of pseudo-accommodation was calculated in each group. In order to determine the possible mechanisms of pseudo-accommodation in children, we compared children with pseudo-accommodation and adults without pseudo-accommodation regarding several parameters: refraction, axial length, corneal topography, aberrometry, pupillary diameter and IOL shift after cyclopentolate instillation. Among the children group, 36 (90xa0%) had pseudo-accommodation versus 2 (7xa0%) in the adult group. We found that spherical equivalent, implant power, corneal multifocality and corneal higher-order aberrations (mainly coma and trefoil) were significantly higher in the pseudo-accommodation group, while pupil diameter and implant shift were not significantly different. Pseudo-accommodation has a high prevalence among non-amblyopic pseudophakic children. Several possible mechanisms have been found to explain pseudo-accommodation in children: a high power of the IOL and a small axial length, maximizing the effect of the IOL shift, corneal multifocality and corneal higher-order aberrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS021533
Cyclopentolate
C17H25NO3