EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Synthesis of Enantiopure Alicyclic Ethers and Their Activity on the Chemosensory Organ of the Ectoparasite of Honey Bees, Varroa destructor.
PMID 27792333
Abstract
The preparation of enantiopure conformationally restricted alicyclic ethers and their inhibitory activities on the chemosensory organ of the Varroa destructor, a parasite of honey bees, are reported in this article. We tested the effect of enantiopure ethers of cis-5-(2'-hydroxyethyl)cyclopent-2-en-1-ol on the Varroa chemosensory organ by electrophysiology, for their ability to inhibit the responses to two honey bee-produced odors that are important for the mite to locate its host: nurse bee head space odor and (E)-β-ocimene, a honey bee brood pheromone. Previous work with the racemic compounds showed that they suppress the mite's olfactory response to its bee host, which led to incorrect host choice. Based on a structure-activity relationship, we predicted that the two most active compounds-cis-1-butoxy-5-(2'-methoxyethyl)cyclopent-2-ene, cy{4,1}, and (cis-1-ethoxy-5-(2'ethoxyethyl)cyclopent-2-ene, cy{2,2}-could have opposite active enantiomers. Here we studied the enantiomers of both ethers, whose preparation involved enzymatic resolution of racemic diol cis-5-(2'-hydroxyethyl)cyclopent-2-en-1-ol using Lipase AK with vinyl acetate. The racemic diol was prepared from commercially available 2,5-norbornadiene. We observed that the responses of the chemosensory organ to honey bee head space volatiles were significantly decreased by both enantiomers of cy{4,1} and cy{2,2}, but that responses to (E)-β-ocimene were decreased significantly only by (+)-cy{4,1} (1R,5S) and (-)-cy{2,2} (1S,5R) and not by their respective enantiomers. The importance of this result is that the racemates could be used to inhibit olfactory detection of bee odors by mites, without a loss in activity relative to the more expensive enantiopure compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B33803
Bicyclo[2.2.1]hepta-2,5-diene, 98%
C7H8