EMAIL THIS PAGE TO A FRIEND
Anticancer research
The Antitumor Effect of Single-domain Antibodies Directed Towards Membrane-associated Catalase and Superoxide Dismutase.
PMID 27793920
Abstract
Neutralizing single-domain antibodies directed towards catalase or superoxide dismutase (SOD) caused efficient reactivation of intercellular reactive oxygen species/reactive nitrogen species (ROS/RNS)-dependent apoptosis-inducing signaling specifically in human tumor cells. Single-domain antibodies targeted tumor cell-specific membrane-associated SOD and catalase, but not the corresponding intracellular enzymes. They were shown to be about 200-fold more effective than corresponding classical recombinant antigen-binding fragments and more than four log steps more efficient than monoclonal antibodies. Combined addition of single-domain antibodies against catalase and SOD caused a remarkable synergistic effect. Proof-of-concept experiments in immunocompromised mice using human tumor xenografts and single-domain antibodies directed towards SOD showed an inhibition of tumor growth. Neutralizing single-domain antibodies directed to catalase and SOD also caused a very strong synergistic effect with the established chemotherapeutic agent taxol, indicating an overlap of signaling pathways. This effect might also be useful in order to avoid unwanted side-effects and to drastically lower the costs for taxol-based therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282332
Benzenesulfonyl fluoride, 99%
C6H5FO2S