EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Dendrimeric Guanidinoneomycin for Cellular Delivery of Bio-macromolecules.
PMID 27806190
Abstract
We present the synthesis of polymeric amino- and guanidinoglycosides prepared by tethering neomycin and guanidinoneomycin to PAMAM dendrimers of generations 2 and 4. The ability of these conjugates to promote cellular uptake of high-molecular-weight cargo is discussed, together with their cytotoxicity and mechanisms of entry. We demonstrate that the presence of multiple guanidinoneomycin carriers on the PAMAM surface plays an important role in promoting cellular uptake of the dendrimers, maintaining the heparan sulfate specificity and negligible cytotoxicity typical of monomeric guanidinoglycoside molecular transporters.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
126209
2,3-Dihydroxybenzoic acid, 99%
C7H6O4