EMAIL THIS PAGE TO A FRIEND
Journal of viral hepatitis
Induction of humoral and cellular immune response to hepatitis B virus (HBV) vaccine can be upregulated by CpG oligonucleotides complexed with Dectin-1 ligand.
PMID 27807909
Abstract
A persistent hepatitis B virus (HBV) infection is characterized by a lack of or a weak immune response to HBV, which may be reflective of tolerance to HBV. Efficient induction of HBV-specific immune response leads to the clearance of HBV in patients with a chronic HBV infection. CpG oligodeoxynucleotides (ODN) has a powerful adjuvant effect in HBV vaccination. A recent report demonstrated that the immunization by B/K CpG ODN (K3) wrapped by the nonagonistic Dectin-1 ligand, schizophyllan (SPG), namely K3-SPG, was more effective in the induction of antigen-specific immune response than that by K3. In this study, we examined the efficacy of K3-SPG as a HBV vaccine adjuvant. Wild-type (WT) mice and HBV transgenic (HBV-Tg) mice were subcutaneously immunized with hepatitis B surface antigen (HBsAg) alone, HBsAg and K3, or HBsAg and K3-SPG. The vaccination with HBsAg and K3-SPG significantly enhanced humoral and cellular immune response to HBV antigen compared to the other vaccinations in WT and HBV-Tg mice. K3-SPG induced the accumulation of dendritic cells (DCs) into draining lymph node and the activation of DCs. The expression of cytokines and chemokines related to Th1 and Th2 responses was upregulated after the vaccination including with K3-SPG. In conclusion, these results indicated that the vaccination using K3-SPG may overcome tolerance even in patients with chronic HBV infection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
703478
trans-2-Bromovinylboronic acid MIDA ester, technical grade
C7H9BBrNO4