EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Design, synthesis, and biological evaluation of structurally modified isoindolinone and quinazolinone derivatives as hedgehog pathway inhibitors.
PMID 27810591
Abstract
The Hedgehog (Hh) signaling pathway is associated with diverse aspects of cellular events, such as cell migration, proliferation, and differentiation throughout embryonic development and tissue patterning. An abnormal Hh signaling pathway is linked to numerous human cancers, including basal cell carcinoma (BCC), medulloblastoma (MB), lung cancer, prostate cancer, and ovarian cancer, and it is therefore a promising target in cancer therapy. Using a structure-hopping approach, we designed new Hh signaling pathway inhibitors with isoindolinone or quinazolinone moieties, which were synthesized and biologically evaluated using an 8xGli-luciferase (Gli-Luc) reporter assay in NIH3T3 cells. Compounds 9-11 and 14 with isoindolinone scaffolds demonstrated moderate Hh inhibitory activity; whereas quinazolinone derivatives 24, 29, 32, 34, and 35 exhibited good potency with submicromolar IC50 values and the analog 28 showed nanomolar IC50 value. Although sonidegib shows a decrease in inhibitory effect on vismodegib resistance-conferring Smo mutants, the structurally modified new compounds not only possess the pharmacophoric properties of Hh pathway inhibition but also preserve the suppressive potency in drug-resistant Smo mutants. Mechanistically, quinazolinone derivatives 28 and 34 suppress Hh signaling by blocking Smo and Gli translocation into the cilia, similar to vismodegib and sonidegib. Additionally, the human microsomal stability of the representative analogs 28 and 34 were determined to be comparable to that of the reference compound sonidegib. Thus, these new scaffolds can serve as a platform for the development of novel cancer therapeutics targeting the Hh pathway.