EMAIL THIS PAGE TO A FRIEND
Neurochemistry international
Microglia derived from the axotomized adult rat facial nucleus uptake glutamate and metabolize it to glutamine inxa0vitro.
PMID 27816478
Abstract
Microglia in the axotomized adult rat facial nucleus (axoFN) have been shown to highly express a glutamate transporter (GLT-1). The microglia appear to serve as glutamate (Glu) scavengers in the axoFN. However, there is no evidence that the microglia actually have the ability to uptake Glu and convert it to Gln. In this study, we investigated whether axoFN-derived microglia (axoFN-microglia) can uptake Glu and metabolize it to Gln. Microglia obtained by explant culture of axoFN on poly(N-isopropylacrylamide)-grafted dishes were non-invasively sub-cultured onto dishes or wells. Immunoblotting and Glu-uptake experiments revealed that the axoFN-microglia uptake (14)C-Glu mainly by GLT-1 activity. Immunoblotting and immunocytochemical methods clarified that axoFN-microglia express the Gln synthetase (GS) protein in the same manner as newborn rat brain-derived primary microglia (NRB-microglia). Biochemical analysis demonstrated that the specific activity of GS of axoFN-microglia is similar to that of NRB-microglia, suggesting that these microglia play equivalent roles in the metabolic conversion of Glu to Gln. Nuclear magnetic resonance analysis clarified that NRB-microglia metabolize [(13)C]Glu to [(13)C]Gln depending on the incubation time, inferring the similar potential of axoFN-microglia. Taken together, these results demonstrate that axoFN-microglia express functional GLT-1 and GS proteins, and are strongly suggested to serve as Glu scavengers inxa0vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1064
Dihydrokainic acid, ≥98% (HPLC), powder
C10H17NO4
214698
Pyridinium dichromate, 98%
C10H10N2 · H2Cr2O7