EMAIL THIS PAGE TO A FRIEND
Oncotarget
Sequential delivery of therapeutic agents using a rationally designed disulfide-linked glycolipid-like nanocarrier.
PMID 27825127
Abstract
Usage of combination therapies to deliver multiple therapeutics to increase treatment efficacy has shown promising results in the clinic. In an effort to maximize the synergistic effect of co-delivery of a drug and siRNA, we have developed a time-dependent sequential drug delivery system (DDS) based on a disulfide-linked chitosan-based nanocarrier (CS-ss-SA) for the co-delivery of paclitaxel (PTX) and Bcl-2 specific siRNA (siBcl-2). This CS-ss-SA nanocarrier is able to transport both drug and siRNA by entrapment of PTX and adsorption of siRNA on the shell by electrostatic attraction. We show that this nanocarrier transports siRNA into tumor cells via its glycolipid-like spatial structure and releases a hydrophobic model drug, Nile Red 8-11 h later. Next, when siRNA and the hydrophobic drug PTX were co-delivered to tumor cells, a synergistic effect was observed in both cell cycle arrest and cell viability. Ultimately, the co-delivery of PTX and siBcl-2 by CS-ss-SA may prove to be more efficacious and may even help overcome drug resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU135281
MISSION® esiRNA, esiRNA human BCL2 (esiRNA1)