EMAIL THIS PAGE TO A FRIEND
Life sciences
Neuroprotective effects of clarithromycin against neuronal damage in cerebral ischemia and in cultured neuronal cells after oxygen-glucose deprivation.
PMID 27825902
Abstract
Rats subjected to transient focal ischemia and cultured neuronal cells subjected to oxygen-glucose deprivation (OGD) were treated with clarithromycin (CAM) to evaluate the effects of CAM in protecting against neuronal damage. Sprague-Dawley rats were subjected to middle cerebral artery occlusion (MCAO) for 90min and then reperfused. Each animal was given an oral dose clarithromycin (CAM, 100mg/kg) or vehicle alone just after the ischemia was commenced. The infarct volume, edema index and neurological performance were assessed after 24 and 72h of reperfusion. The cerebral blood flow (CBF) was measured with an MRI system at 90min after MCAO. After 24 and 72h, oxidative stress (4-HNE, 8-OHdG) and inflammation (Iba-1, TNF-α) were assessed by immunohistochemical analyses and degenerative cells were assessed in the cortex by Fluoro-Jade C (FJC) labeling. The cultured neuronal cells were also used to examine the effects of CAM exposure on the viability of the cells after OGD. CBF was unchanged between the two groups. Significant reductions of the infarct volume and edema index, an improved neurological deficit score, a significant suppression of 4-HNE and 8-OHdG expression, marked reductions of Iba-1 and TNF-α expression, and a significant reduction of FJC-positive cells were also observed in the CAM-treated animals at both time points. Treatment with 10μM and 100μM CAM in vitro significantly reduced cell death after OGD. CAM appears to provide antioxidant and anti-inflammatory effects and protect against neuronal damage after cerebral ischemia and OGD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46043
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)
C46H50Cl4N8