EMAIL THIS PAGE TO A FRIEND
Optics express
Tracking E. coli runs and tumbles with scattering solutions and digital holographic microscopy.
PMID 27828208
Abstract
We use in-line digital holographic microscopy to image freely swimming E. coli. We show that fitting a light scattering model to E. coli holograms can yield quantitative information about the bacterium's body rotation and tumbles, offering a precise way to track fine details of bacterial motility. We are able to extract the cell's three-dimensional (3D) position and orientation and recover behavior such as body angle rotation during runs, tumbles, and pole reversal. Our technique is label-free and capable of frame rates limited only by the camera.