EMAIL THIS PAGE TO A FRIEND
BMC biotechnology
Repair of cartilage defects in osteoarthritis rats with induced pluripotent stem cell derived chondrocytes.
PMID 27829414
Abstract
The incapacity of articular cartilage (AC) for self-repair after damage ultimately leads to the development of osteoarthritis. Stem cell-based therapy has been proposed for the treatment of osteoarthritis (OA) and induced pluripotent stem cells (iPSCs) are becoming a promising stem cell source. Three steps were developed to differentiate human iPSCs into chondrocytes which were transplanted into rat OA models induced by monosodium iodoacetate (MIA). After 6xa0days embryonic body (EB) formation and 2xa0weeks differentiation, the gene and protein expression of Col2A1, GAG and Sox9 has significantly increased compare to undifferentiated hiPSCs. After 15xa0weeks transplantation, no immune responses were observed, micro-CT showed gradual engraftment and the improvement of subchondrol plate integrity, and histological examinations demonstrated articular cartilage matrix production. hiPSC could be an efficient and clinically translatable approach for cartilage tissue regeneration in OA cartilages.