EMAIL THIS PAGE TO A FRIEND
Current microbiology
Identification of a Halotolerant Mutant via In Vitro Mutagenesis in the Cyanobacterium Fremyella diplosiphon.
PMID 27844126
Abstract
Energy metabolism and photosynthetic pigment accumulation are affected by salt stress in cyanobacteria leading to cessation of growth. In this study, the effect of salinity on the freshwater cyanobacterium, Fremyella diplosiphon, was investigated and mutagenesis-based efforts were undertaken to enhance salt tolerance. Salinity at a concentration of 10xa0g/L sodium chloride (NaCl) inhibited growth of wildxa0type F. diplosiphon under white, red, and green light. Efforts to enhance halotolerance resulted in a mutant that could survive in 20xa0g/L NaCl for 15 generations with no significant reduction in phycobiliproteins (phycocyanin, phycoerythrin, and allophycocyanin) or chlorophyll a. Gene expression measured by quantitative reverse transcription-polymerase chain reaction revealed a three-fold increase in tripartite ATP-independent periplasmic transporters (TRAP) solute receptor transcript in the mutant compared to wild type. Our discovery of a TRAP transporter system in F. diplosiphon and its possible role in salinity response enables growth in brackish waters, which enhances its potential for biotechnological applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7152
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
C81H118N20O23