EMAIL THIS PAGE TO A FRIEND
Scientific reports
Insulin-IGF signaling affects cell transformation in the BALB/c 3T3 cell model.
PMID 27849005
Abstract
The increased cancer mortality of diabetes type 2 patients is most likely an evidence of the tight connection between tumor development and energy metabolism. A major focus of today's research is still the identification of key proteins of both diseases and the development of corresponding inhibitors. In this study we combined the two-stage BALB/c-3T3 cell transformation assay (BALB-CTA) with the IR/IGF-1R inhibitor OSI-906 (linsitinib) and analyzed alterations in protein activity and energy parameters in non-transformed as well as transformed cells. OSI-906 successfully inhibited the phosphorylation of IR/IGF-1R and decreased cell growth in non-transformed cells. In the BALB-CTA, a permanent treatment with OSI-906 reduced cellular transformation dose-dependently, whereas a temporary treatment gave evidence for a preventive effect in the promotion phase. Furthermore, even though several key proteins were affected, it was possible to show that the phosphorylation of GSK3, Erk 1/2 and the S6 protein are not crucial for the cell foci reducing effect of OSI-906. Taken together, the BALB-CTA confirmed results of OSI-906 from animal studies and enhanced the knowledge of its mode of action. Therefore, the BALB-CTA offers the opportunity to analyze alterations in the transformation process more precisely and will be helpful to identify effective cancer treatments.