EMAIL THIS PAGE TO A FRIEND
Oncotarget
Isochaihulactone-induced DDIT3 causes ER stress-PERK independent apoptosis in glioblastoma multiforme cells.
PMID 27852055
Abstract
The endoplasmic reticulum (ER) is a major site of cellular homeostasis regulation. Under the ER stress condition, Glioblastoma multiform (GBM) cells activate the unfolded protein response. In this study, we discovered isochaihulactone, a natural compound extracted from the Chinese traditional herb Nan-Chai-Hu, which can disrupt ER homeostasis in GBM cell lines. It can induce DNA damage inducible transcript 3 (DDIT3) expression which is independent of 78 kDa glucose-regulated protein (GRP78) and protein kinase RNA-like endoplasmic reticulum kinase (PERK) expression. Flow cytometry results revealed that isochaihulactone trigger the cell cycle arrest at G2/M phase and apoptosis in GBM cells. Isochaihulactone induced DDIT3 led to the expression of NAG-1. The in vivo study showed that isochaihulactone suppressed tumor growth, and DDIT3 and Caspase3 overexpressed in the xenograft model, which is consistent with the in vitro study. Overall, the data revealed that isochaihulactone disrupted ER homeostasis in cancer cells by increasing DDIT3 and NAG-1 expression. Our finding also provides a therapeutic strategy by using isochaihulactone for GBM treatment.