EMAIL THIS PAGE TO A FRIEND
Oncotarget
Overexpression of FZD7 promotes glioma cell proliferation by upregulating TAZ.
PMID 27852064
Abstract
Gliomas are the most prevalent type of primary brain tumors in adults, accounting for more than 40% of neoplasm in the central nervous system. Frizzled-7 (FZD7) is a seven-pass trans-membrane Wnt receptor that plays a critical role in the development of various tumors. In this study, we detected high-level FZD7 expression in glioma and its overexpression was associated with advanced tumor stage. In vitro functional assays showed that forced overexpression of FZD7 promoted proliferation of gliomas cells, whereas knockdown of endogenous FZD7 significantly suppressed proliferation ability of these cells. In a xenograft assay, FZD7 was also found to promote the growth of glioma cells. We further found that FZD7 could activate transcriptional coactivator with PDZ-binding motif (TAZ), and TAZ was required for FZD7 to promote cell proliferation in glioma. Furthermore, the univariate analysis of survival shows that glioma patients with high FZD7 expression have a shorter survival. In conclusion, our findings demonstrate that FZD7 may promote glioma cell proliferation via upregulation of TAZ.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
685720
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
C30H27N3