EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
MiR-33a suppresses proliferation of NSCLC cells via targeting METTL3 mRNA.
PMID 27856248
Abstract
Methyltransferase like 3 (METTL3) was incipiently known as a methyltransferase which was responsible for N(6)-methyladenosine (m(6)A) methylation. METTL3 can promote the expression of several crucial oncoproteins and its high expression enhanced proliferation, survival, and invasion of human lung cancer cells. However, how METTL3 was regulated is seldom understood in non-small-cell lung carcinoma (NSCLC). In the present study, miR-33a was suspicious to target to the 3'-untranslated region (3'UTR) of METTL3 mRNA via in silico prediction. Besides, the expressions of METTL3 were higher in NSCLC tissues than those in adjacent tissues, and METTL3 expressions were positively related to the expressions of miR-33a in NSCLC tissues which confirmed by quantitative real-time polymerase chain reaction (qRT-PCR). MiR-33a can directly target to the 3'UTR of METTL3 mRNA which examined by luciferase reporter gene assay. Moreover, we found that miR-33a can reduce the expression of METTL3 at both mRNA and protein levels using reverse transcription-polymerase chain reaction (RT-PCR) and Western blot analysis. Functionally, miR-33a can reduce the proliferation of A549 and NCI-H460xa0cells. Conversely, inhibition of miR-33a by anti-miR-33a can rescue that using 4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2-H-tetrazolium bromide (MTT) and 5-ethynyl-2-deoxyuridine (EdU) assay. Similarly, miR-33a can reduce cellular anchorage-independent growth of A549xa0cells. Additionally, the negative influences of miR-33a on the downstream genes of METTL3 were examined by Western blot analysis. Thus, we concluded that miR-33a can attenuate NSCLC cells proliferation via targeting to the 3'UTR of METTL3 mRNA. Our findings provide new insights into the mechanism of METTL3 regulation by micro RNA, and supports METTL3 as a therapeutic target in NSCLC.