EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Pulsed electromagnetic field inhibits RANKL-dependent osteoclastic differentiation in RAW264.7 cells through the Ca(2+)-calcineurin-NFATc1 signaling pathway.
PMID 27856256
Abstract
Pulsed electromagnetic field (PEMF) has been reported to improve bone healing in osteoporosis patients. However, the precise mechanism has remained largely unknown. This study aimed to investigate the effects of PEMF on nuclear factor κB ligand (RANKL)-dependent osteoclastic differentiation and the Ca(2+)-calcineurin-NFATc1 signaling pathway in RAW264.7xa0cells inxa0vitro. Treating RAW264.7xa0cells with RANKL for 4 days induced osteoclastic differentiation inxa0vitro, and the formation of multinucleated osteoclasts, bone resorption-pit formation, tartrate-resistant acid phosphatase (TRAP) activity and the protein levels of cathepsin K, TRAP, Nuclear factor of activated T-cells, cytoplasmic 1 (NFATc1) and matrix metalloproteinase 9 (MMP-9) were significantly decreased. The mRNA levels of specific genes related to osteoclastogenesis (TRAP, NFATc1, CTSK and MMP-9) were also reduced. Moreover, the oscillations of intracellular Ca(2+) in RANKL-dependent RAW264.7xa0cells were suppressed by PEMF, as well as by inhibitors of membrane and store-operated Ca(2+) channels. Meanwhile, calcineurin activity was increased, although its protein level was not changed. PEMF increased phospho-NFATc1 in the cytosol while suppressing the nuclear translocation of NFATc1, thus inhibiting osteoclastic differentiation by suppressing the Ca(2+)-calcineurin-NFATc1 signaling pathway. Although many questions remain unresolved, to our knowledge, this is the first report demonstrating that PEMF is beneficial against RANKL-dependent osteoclastic differentiation in RAW264.7xa0cells inxa0vitro via inhibiting the Ca(2+)-calcineurin-NFATc1 signaling pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7152
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
C81H118N20O23