EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Development of a bacterial-based negative selection system for rapid screening of active single guide RNAs.
PMID 27858321
Abstract
To develop an in vitro method for rapid evaluation of the capability of a designed single guide RNAs (sgRNAs) to guide Cas9 nucleases to cleave target loci in mammalian cells. We constructed a Cas9/sgRNA plasmid with two SP6 promoters to simultaneously express Cas9 nuclease and the sgRNA and a negative selection plasmid harbouring a target site of the sgRNA. After co-transforming chemically competent E. coli DH5α cells with the two plasmids, the transformants were plated at a low density on two LB plates: one containing only ampicillin and the other containing both ampicillin and chloramphenicol. The colony-count on the ampicillinxa0+xa0chloramphenicol plate was compared with that on the ampicillin-only plate to calculate the survival percentage. The survival % was negatively correlated with the genome editing efficiency of the sgRNA in mammalian cells evaluated by a T7 endonuclease 1 (T7E1) assay (r ranged from -0.8 to -0.92). This system eliminates the need for cell culture, transfection, FACS sorting, PCR and T7E1 nuclease treatment, and significantly reduces the cost of screening for active sgRNAs, especially in the case of large-scale screening. We have developed a bacterial-based negative selection system for rapid screening of active sgRNAs in mammalian cells at a very low cost.