EMAIL THIS PAGE TO A FRIEND
Water research
Removal characteristics of DON in pharmaceutical wastewater and its influence on the N-nitrosodimethylamine formation potential and acute toxicity of DOM.
PMID 27871050
Abstract
Previous research has focused on dissolved organic carbon (DOC) as a surrogate for dissolved organic matter (DOM) in pharmaceutical wastewater. Dissolved organic nitrogen (DON) as a part of DOM has received little attention. This study investigated the removal characteristics of DON and its influence on the N-nitrosodimethylamine formation potential (NDMA FP) and acute toxicity of DOM in a full-scale hydrolysis/acidificationxa0+xa0anaerobic/anoxic/aerobicxa0+xa0moving bed biofilm reactor (MBBR) process treating pharmaceutical wastewater. Results showed that maximum removal of DON (68xa0±xa012%) was present in the anaerobic process. The removal of DON by anoxic and aerobic processes was negligible as a result of the production of new N-containing compounds that are characteristic of proteins/amino sugars and lipids. DON concentration decreased significantly in the MBBR process (pxa0