EMAIL THIS PAGE TO A FRIEND
Plant biotechnology journal
Generation of targeted mutant rice using a CRISPR-Cpf1 system.
PMID 27875019
Abstract
CRISPR-Cpf1 is a newly identified CRISPR-Cas system, and Cpf1 was recently engineered as a molecular tool for targeted genome editing in mammalian cells. To test whether the engineered CRISPR-Cpf1 system could induce the production of rice mutants, we selected two genome targets in the OsPDS and OsBEL genes. Our results show that both targets could be efficiently mutated in transgenic rice plants using CRISPR-Cpf1. We found that pre-crRNAs with a full-length direct repeat sequence exhibited considerably increased efficiencies compared with mature crRNAs. In addition, the specificity and transmission of the mutation were investigated, and the behaviours of crRNA-Cpf1-induced plant targeted genome mutagenesis were assessed. Taken together, our results indicate that CRISPR-Cpf1 expression via stable transformation can efficiently generate specific and heritable targeted mutations in rice and thereby constitutes a novel and important approach to specific and precise plant genome editing.