EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Hydrins, hydroosmotic neurohypophysial peptides: osmoregulatory adaptation in amphibians through vasotocin precursor processing.
PMID 2787509
Abstract
From neurointermediate pituitary glands of Xenopus laevis and Rana esculenta, previously unreported peptides termed hydrins, active on water permeability of frog urinary bladder and frog skin (Brunn or "water-balance" effect), have been isolated and sequenced. These peptides seem to be derived from the pro-vasotocin-neurophysin precursor. Hydrin 1, found in Xenopus, has been identified as vasotocin C-terminally extended with the Gly-Lys-Arg sequence; hydrin 2, found in Rana, has been identified as vasotocin C-terminally extended with glycine. Hydrin 2 has been detected in several Ranidae (R. esculenta, Rana temporaria, Rana pipiens) and Bufonidae (Bufo bufo, Bufo ictericus) and appears to have a large distribution in terrestrial or semiaquatic anurans. Hydrins, in contrast to vasotocin, are not active on rat uterus or rat blood pressure. They are absent from other vasotocin-bearers such as birds and could be involved specifically in water-electrolyte regulation of amphibians.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V7009
[Arg8]-Vasotocin-Gly-Lys, ≥97% (HPLC)
C51H81N17O15S2
V1258
[Arg8]-Vasotocin-Gly-Lys-Arg, ≥97% (HPLC)
C57H93N21O16S2