EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Involvement of bleomycin hydrolase and poly(ADP-ribose) polymerase-1 in Ubc9-mediated resistance to chemotherapy agents.
PMID 27878232
Abstract
Ubiquitin-conjugating protein 9 (Ubc9), the sole enzyme for sumoylation, plays critical roles in many physiological functions, such as DNA damage repair and genome integrity. Its overexpression led to poor prognosis and drug resistance in tumor chemotherapy. However, the underlying mechanism by which Ubc9 promotes tumor progress and influences the susceptibility to antitumor agents remains elusive. In this study, we used nine antitumor agents with distinct actions to explore Ubc9-mediated resistance in human breast carcinoma MCF-7 cells. Increase of susceptibility, respectively, to boningmycin, hydroxycamptothecine, cis-dichlorodiamineplatinum, 5-fluorouracil, vepeside and gemcitabine, but not for doxorubicin, vincristine and norcantharidin, was observed after the knockdown of Ubc9 protein level with RNA interference. Reduction of bleomycin hydrolase and poly(ADP-ribose) polymerase-1 levels after knockdown of Ubc9 suggests their contribution to Ubc9-mediated drug resistance. This is the first report on the sensitivity to hydroxycamptothecine, cis-dichlorodiamineplatinum and gemcitabine that increased after knockdown of bleomycin hydrolase at protein level. In conclusion, Ubc9 plays different roles of action in antitumor agents in chemotherapy. The process requires bleomycin hydrolase and poly(ADP-ribose) polymerase-1. The results are beneficial to deeply understanding of Ubc9 functions and for precise prediction of chemotherapy outcomes in tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8784
Norcantharidin, solid
C8H8O4