EMAIL THIS PAGE TO A FRIEND
Molecular and cellular endocrinology
Functional characterization of two naturally occurring mutations V(221)G and T(449)N in the follicle stimulating hormone receptor.
PMID 27889471
Abstract
Naturally occurring mutations in follicle stimulating hormone receptor (FSHR) affect the receptor function. Here, we characterized two such previously reported mutations, V(221)G and T(449)N, in the extracellular domain and transmembrane helix 3, of FSHR, respectively. Functional studies with the V(221)G mutant demonstrated an impairment in FSH binding and signaling. Validation of X-ray crystallography data indicating the contribution of FSHR specific residues in the vicinity of V(221) to contribute to FSH-FSHR interaction was carried out. Inxa0vitro mutational studies showed that these residues are determinants of both FSH binding and FSH induced signaling. Analysis of the T(449)N mutation revealed that it results in an increase in FSH binding and high cAMP response at lower doses of FSH. A marginal hCG induced and no TSH induced cAMP production was also observed. These findings corroborated with the clinical manifestations of primary amenorrhea (V(221)G) and spontaneous ovarian hyperstimulation syndrome (T(449)N) in women harbouring these mutations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323616
3′-Deoxythymidine, 98%
C10H14N2O4