EMAIL THIS PAGE TO A FRIEND
Scientific reports
Novel Ti-Ta-Hf-Zr alloys with promising mechanical properties for prospective stent applications.
PMID 27897215
Abstract
Titanium alloys are receiving increasing research interest for the development of metallic stent materials due to their excellent biocompatibility, corrosion resistance, non-magnetism and radiopacity. In this study, a new series of Ti-Ta-Hf-Zr (TTHZ) alloys including Ti-37Ta-26Hf-13Zr, Ti-40Ta-22Hf-11.7Zr and Ti-45Ta-18.4Hf-10Zr (wt.%) were designed using the d-electron theory combined with electron to atom ratio (e/a) and molybdenum equivalence (Moeq) approaches. The microstructure of the TTHZ alloys were investigated using optical microscopy, XRD, SEM and TEM and the mechanical properties were tested using a Vickers micro-indenter, compression and tensile testing machines. The cytocompatibility of the alloys was assessed using osteoblast-like cells in vitro. The as-cast TTHZ alloys consisted of primarily β and ω nanoparticles and their tensile strength, yield strength, Young's modulus and elastic admissible strain were measured as being between 1000.7-1172.8 MPa, 1000.7-1132.2 MPa, 71.7-79.1 GPa and 1.32-1.58%, respectively. The compressive yield strength of the as-cast alloys ranged from 1137.0 to 1158.0 MPa. The TTHZ alloys exhibited excellent cytocompatibility as indicated by their high cell viability ratios, which were close to that of CP-Ti. The TTHZ alloys can be anticipated to be promising metallic stent materials by virtue of the unique combination of extraordinarily high elastic admissible strain, high mechanical strength and excellent biocompatibility.