EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Oxidative Stress Mediated Hippocampal Neuron Apoptosis Participated in Carbon Disulfide-Induced Rats Cognitive Dysfunction.
PMID 27900598
Abstract
Occupational exposure to carbon disulfide (CS2) exhibits central nervous systems toxicity. But the mechanism is unclear. The present study was designed to investigate the relationship between the CNS damage and cognitive dysfunction caused by CS2, and eventually reveal the possible oxidative-related mechanism of hippocampus pathological changes in CS2 exposed rats. Male Wistar rats were administrated with CS2 at dosage of 200, 400 and 600xa0mg/kg for consecutive 20xa0days, respectively. Cognitive performances were evaluated by Morris water maze tests. Thionin and immunohistochemical analysis were used to investigate the hippocampal neuron damage, and the expression of apoptosis related proteins (cleaved-caspase 3, Bax and Bcl-2) were detected to explore the possible mechanisms of neuronal loss. Oxidative stress parameters were checked by commercial assay kits. Rats exposed to CS2 displayed cognitive dysfunction manifested as decreased spatial learning ability and memory lesion. Pathological changes and significant neuron loss were observed in hippocampus, especially in CA1 and CA3 sub-regions. Mitochondria-dependent apoptosis pathway was implicated in the CS2-induced neuronal loss which was demonstrated by the up-regulation of cleaved-caspase 3 and Bax accompanied with down-regulation of Bcl-2. Furthermore, extensive oxidative stress induced by CS2 was also revealed by the measurement of ROS, RNS, MDA, GSH&GSSG and antioxidant enzymes (CAT, T-SOD, and GSH-Px). Our study suggested that oxidative stress mediated hippocampal neuron apoptosis might play an important role in CS2 induced CNS damage and cognitive dysfunction.