EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Metabolic Changes in the Bilateral Visual Cortex of the Monocular Blind Macaque: A Multi-Voxel Proton Magnetic Resonance Spectroscopy Study.
PMID 27909856
Abstract
The metabolic changes accompanied with adaptive plasticity in the visual cortex after early monocular visual loss were unclear. In this study, we detected the metabolic changes in bilateral visual cortex of normal (group A) and monocular blind macaque (group B) for studying the adaptive plasticity using multi-voxel proton magnetic resonance spectroscopy ((1)H-MRS) at 32xa0months after right optic nerve transection. Then, we compared the N-Acetyl aspartate (NAA)/Creatine (Cr), myoinositol (Ins)/Cr, choline (Cho)/Cr and Glx (Glutamate + glutamine)/Cr ratios in the visual cortex between two groups, as well as between the left and right visual cortex of group A and B. Compared with group A, in the bilateral visual cortex, a decreased NAA/Cr and Glx/Cr ratios in group B were found, which was more clearly in the right visual cortex; whereas the Ins/Cr and Cho/Cr ratios of group B were increased. All of these findings were further confirmed by immunohistochemical staining. In conclusion, the difference of metabolic ratios can be detected by multi-voxel (1)H-MRS in the visual cortex between groups A and B, which was valuable for investigating the adaptive plasticity of monocular blind macaque.
Related Materials