EMAIL THIS PAGE TO A FRIEND
Cancer letters
NF-kB-regulated exosomal miR-155 promotes the inflammation associated with arsenite carcinogenesis.
PMID 27913196
Abstract
In the cancer microenvironment, extracellular communication allows various types of cells to coordinate and execute biological functions. Emerging evidence indicates that exosomes, as mediators of cell communication, are involved in tumor progression. Little is known, however, about the mechanism by which exosomal miRNAs regulate inflammatory infiltration in arsenite-induced liver cancer. The present research aimed to determine if miRNAs secreted from arsenite-transformed human hepatic epithelial (L-02) cells are transferred into normal L-02 and THLE-3 cells, which are functionally active in the recipient cells. The results show that the exosomes from arsenite-transformed L-02 cells enhance miR-155 expression and the pro-inflammatory properties of normal L-02 and THLE-3 cells. Transformed cells transfer miR-155 into normal L-02 cells via exosomes. The inhibition of NF-κB by siRNA and inhibitor, which reduces miR-155 levels in exosomes derived from transformed L-02 cells, blocks inflammation. Arsenite-transformed cells secrete exosomes to enhance inflammation, but the inhibition of the synthesis of exosomes fails to stimulate inflammation. miR-155 is involved in exosome-mediated intercellular communication between neoplastic and normal liver cells. In addition, miR-155, IL-6, and IL-8 were over-expressed in the serum of arsenite exposure group. And there was a positive correlation between miR-155 and IL-6 or IL-8 levels. Further, exosomal miR-155 was up-regulated in the serum of arsenite exposure group. Thus, these results show that exosomes derived from transformed L-02 cells transfer miR-155 to surrounding cells, which induces pro-inflammatory activity of normal liver cells. The findings support the concept that exosomal miRNAs are involved in cell-cell communication during carcinogenesis induced by environmental chemicals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU141461 MISSION® esiRNA, esiRNA human RELA (esiRNA1)