EMAIL THIS PAGE TO A FRIEND
Molecular ecology
Multiple processes drive genetic structure of humpback whale (Megaptera novaeangliae) populations across spatial scales.
PMID 27914203
Abstract
Elucidating patterns of population structure for species with complex life histories, and disentangling the processes driving such patterns, remains a significant analytical challenge. Humpback whale (Megaptera novaeangliae) populations display complex genetic structures that have not been fully resolved at all spatial scales. We generated a data set of nuclear markers for 3575 samples spanning the seven breeding stocks and substocks found in the South Atlantic and western and northern Indian Oceans. For the total sample, and males and females separately, we assessed genetic diversity, tested for genetic differentiation between putative populations and isolation by distance, estimated the number of genetic clusters without a priori population information and estimated rates of gene flow using maximum-likelihood and Bayesian approaches. At the ocean basin scale, structure is governed by geographical distance (IBD Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0662
6-Carboxyfluorescein, ~97% (HPLC)
C21H12O7
80666
Hexarelin, ≥90% (HPLC)
C47H58N12O6