EMAIL THIS PAGE TO A FRIEND
Alcohol (Fayetteville, N.Y.)
Moderate alcohol consumption after a mental stressor attenuates the endocrine stress response.
PMID 27916140
Abstract
Alcohol is often consumed to reduce tension and improve mood when exposed to stressful situations. Previous studies showed that moderate alcohol consumption may reduce stress when alcohol is consumed prior to a stressor, but data on the effect of alcohol consumption after a mental stressor is limited. Therefore, our objective was to study whether moderate alcohol consumption immediately after a mental stressor attenuates the stress response. Twenty-four healthy men (age 21-40xa0y, BMI 18-27xa0kg/m(2)) participated in a placebo-controlled trial. They randomly consumed 2 cans (660xa0mL, ∼26xa0g alcohol) of beer or alcohol-free beer immediately after a mental stressor (Stroop task and Trier Social Stress Test). Physiological and immunological stress response was measured by monitoring heart rate and repeated measures of the hypothalamic-pituitary-adrenal axis (HPA-axis), white blood cells and a set of cytokines. After a mental stressor, cortisol and adrenocorticotropic hormone (ACTH) concentrations were 100% and 176% more reduced at 60xa0min (Pxa0=xa00.012 and Pxa0=xa00.001, respectively) and 92% and 60% more reduced at 90xa0min (Pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4000
trans-Dehydroandrosterone, ≥99%
C19H28O2