EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and pharmacology
The injury of fine particulate matter from cooking oil fumes on umbilical cord blood vessels in vitro.
PMID 27918956
Abstract
Cooking oil fumes (COFs) derived PM2.5 is the major source of indoor air pollution in Asia. For this, a pregnant rat model within different doses of cooking oil fumes (COFs) derived PM2.5 was established in pregnancy in our research. Our previous studies have showed that exposure to COFs-derived PM2.5 was related to adverse pregnancy outcomes. However, the mechanisms of signaling pathways remain unknown. Therefore, this study aimed to investigate the underlying mechanisms induced by COFs-derived PM2.5 injury on umbilical cord blood vessels (UCs) in vitro. Exposure to COFs-derived PM2.5 resulted in changing the expression of eNOS, ET-1, ETRA, and ETRB. In additions, western blot analysis indicated that the HIF-1α/iNOS/NO signaling pathway and VEGF/VEGFR1/iNOS signaling pathway were involved in UCs injury triggered by COFs-derived PM2.5. In conclusion, our data suggested that exposure to COFs-derived PM2.5 resulted in increasing of oxidative stress and inflammation, as well as dysfunction of UCs.