EMAIL THIS PAGE TO A FRIEND
Anaerobe
Rhodomyrtone inhibits lipase production, biofilm formation, and disorganizes established biofilm in Propionibacterium acnes.
PMID 27923605
Abstract
Virulence enzymes and biofilm a play crucial role in the pathogenesis of Propionibacterium acnes, a major causative agent of acne vulgaris. In the present study, the effects of rhodomyrtone, a pure compound identified from Rhodomyrtus tomentosa (Aiton) Hassk. leaves extract against enzyme production and biofilm formation production by 5 clinical isolates and a reference strain were evaluated. The degree of hydrolysis by both lipase and protease enzymes significantly decreased upon treatment with the compound at 0.125-0.25xa0μg/mL (pxa0
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W222305
Tributyrin, 97%, FG
C15H26O6