EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Megastigmane glycosides from leaves of Eucommia ulmoides Oliver with ACE inhibitory activity.
PMID 27923676
Abstract
Four new megastigmane glycosides, eucomegastigsides A-D (2, 3, 5 and 7), together with three known megastigmane glycosides, (6R, 7E, 9R)-9-hydroxy-4, 7-megastigmadien-3-one-9-O-[α-l-arabinopyranosyl-(l→6)-β-d-glucopyranoside (1), foliasalacioside B1 (4) and eleganoside A (6), were isolated from the leaves of Eucommia ulmoides Oliver. Their anti-hypertensive effect was investigated in vitro based on the inhibition of Angiotensin Converting Enzyme (ACE) using HPLC. The results showed that the isolates (2, 3, 4, 5, 7) had moderate inhibitory effects on ACE in vitro compared with captopril.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56395
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)
C10H17NO3