EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
B cell antigen extraction is regulated by physical properties of antigen-presenting cells.
PMID 27923880
Abstract
Antibody production and affinity maturation are driven by B cell extraction and internalization of antigen from immune synapses. However, the extraction mechanism remains poorly understood. Here we develop DNA-based nanosensors to interrogate two previously proposed mechanisms, enzymatic liberation and mechanical force. Using antigens presented by either artificial substrates or live cells, we show that B cells primarily use force-dependent extraction and resort to enzymatic liberation only if mechanical forces fail to retrieve antigen. The use of mechanical forces renders antigen extraction sensitive to the physical properties of the presenting cells. We show that follicular dendritic cells are stiff cells that promote strong B cell pulling forces and stringent affinity discrimination. In contrast, dendritic cells are soft and promote acquisition of low-affinity antigens through low forces. Thus, the mechanical properties of B cell synapses regulate antigen extraction, suggesting that distinct properties of presenting cells support different stages of B cell responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219924
4-Hydroxy-3-nitrophenylacetic acid, 99%
C8H7NO5