EMAIL THIS PAGE TO A FRIEND
Nanoscale
Nanoconfined self-assembly on a grafted graphitic surface under electrochemical control.
PMID 27924342
Abstract
Highly oriented pyrolytic graphite (HOPG) can be covalently grafted with aryl radicals generated via the electrochemical reduction of 3,5-bis-tert-butyl-diazonium cations (3,5-TBD). The structure of the grafted layer and its stability under electrochemical conditions were assessed with electrochemical scanning tunneling microscopy (EC-STM) and cyclic voltammetry (CV). Stable within a wide (>2.5 V) electrochemical window, the grafted species can be locally removed using EC-STM-tip nanolithography. Using dibenzyl viologen as an example, we show that the generated nanocorrals of bare graphitic surface can be used to study nucleation and growth of self-assembled structures under conditions of nanoconfinement and electrochemical potential control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
273104
3,5-Di-tert-butylaniline, 98%
C14H23N