EMAIL THIS PAGE TO A FRIEND
Anais da Academia Brasileira de Ciencias
Production of crispy bread snacks containing chicken meat and chicken meat powder.
PMID 27925030
Abstract
Chicken meat in two different forms (chicken meat and chicken meat powder) were added into white flour and whole wheat blend baguette bread formulations for protein enrichment and finally developing new and healthy snacks. The chicken meat and powder levels were 10% for white flour baguette, and 15% for whole wheat blend. The dried baguette samples were packaged under 100% N2, and physical, chemical, microbiological and sensorial properties were evaluated during 3 months of storage. Protein content of chicken meat powder added samples were found statistically higher than chicken meat added samples. Hardness of the snacks was significantly affected from type of chicken meat, such as values were higher for chicken meat added samples than chicken meat powder added samples. Lipid oxidation of the snacks was determined by TBA analysis, and TBA value for whole wheat mixture snack with 15% of chicken meat was the highest among all during storage. The highest overall acceptance score was obtained from white flour snack with 10% chicken meat. There was no coliform bacteria detected during storage and the results of yeast-mold count and aerobic plate count of snacks remained between the quantitative ranges.