EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Bone marrow mesenchymal stem cells and their conditioned media could potentially ameliorate ovalbumin-induced asthmatic changes.
PMID 27930984
Abstract
The major feature of asthma is governed by chronic airway inflammation. This investigation was proposed to achieve the suitable candidate for ameliorating long-term chronic asthmatic changes of respiratory tract. 36 rats were classified into healthy (C) and ovalbumin (OVA)-sensitized animals (S). To sensitize, the rats were exposed to OVA over a course of 32±1days. One day after sensitization, equal six different groups were subjected to experimental procedure (n=6); Rats only received intratracheally 50ml PBS (CPT and SPT groups), 50μl conditioned medium (CM) (CST and SST groups) and 50μl PBS containing 2×10(6) rat bone marrow-derived mesenchymal stem cells (rBMMSCs) (CCT and SCT groups). Two weeks after treatment, tracheal responsiveness, immunologic responses and recruitment of rBMMSCs into the lung as well as pathological changes were evaluated. A high degree of tracheal responsiveness, total white blood cell and percentages of eosinophil and neutrophil was significantly recorded in all sensitized groups rather than of controls (p<0.001 to p<0.05). Of interest, all above-mentioned parameters decreased significantly in SST and notably SCT groups as compared to S group (p<0.001 to p<0.05). The results revealed decrease number of blood CD3(+)CD4(+) and concurrent increase in CD3(+)CD8(+) in all sensitized rats as compared to control (p<0.001 to p<0.05). Noticeably, no significant modulatory effects of either cell or CM administration were achieved on the CD3(+)CD4(+) and CD3(+)CD8(+) populations in non-asthmatic rats. Moreover, the number of CD3(+)CD4(+) in SST and SCT groups tended to increase, which coincided with a decreased manner of CD3(+)CD8(+) populations as compared with S group (p<0.001 to p<0.05). However, the CD3(+)CD4(+) cells in SCT rats were significantly higher than the group SST (p<0.01) whereas CD3(+)CD8(+) cells diminished simultaneously (p<0.001). Real-time PCR analysis further showed that both CM and particularly MSCs changed the expression of interleukin (IL)-4 and IL-10 in the asthmatic groups to the near level of control rats (p<0.001 to p<0.05). Histopathological analysis revealed a profound reduction of lungs injuries in asthmatic rats when received CM and peculiarly mesenchymal stem cells (p<0.01 to p<0.05). Our study shed light on the superior effects of rBMMSCs, rather than CM, in attenuating of chronic asthmatic changes in the rat model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C102
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
C12H16N4O2