EMAIL THIS PAGE TO A FRIEND
Genome
Development of oligonucleotides and multiplex probes for quick and accurate identification of wheat and Thinopyrum bessarabicum chromosomes.
PMID 27936984
Abstract
In comparison with general FISH for preparing probes in terms of time and cost, synthesized oligonucleotide (oligo hereafter) probes for FISH have many advantages such as ease of design, synthesis, and labeling. Low cost and high sensitivity and resolution of oligo probes greatly simplify the FISH procedure as a simple, fast, and efficient method of chromosome identification. In this study, we developed new oligo and oligo multiplex probes to accurately and efficiently distinguish wheat (Triticum aestivum, 2n = 6x, AABBDD) and Thinopyrum bessarabicum (2n = 2x = 14, JJ) chromosomes. The oligo probes contained more nucleotides or more repeat units that produced stronger signals for more efficient chromosome painting. Four Th. bessarabicum-specific oligo probes were developed based on genomic DNA sequences of Th. bessarabicum chromosome arm 4JL, and one of them (oligo DP4J27982) was pooled with the oligo multiplex #1 to simultaneously detect wheat and Th. bessarabicum chromosomes for quick and accurate identification of Chinese Spring (CS) - Th. bessarabicum alien chromosome introgression lines. Oligo multiplex #4 revealed chromosome variations among CS and eight wheat cultivars by a single round of FISH analysis. This research demonstrated the high efficiency of using oligos and oligo multiplexes in chromosome identification and manipulation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3880
N-(3-Fluoranthyl)maleimide
C20H11NO2