EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological and toxicological methods
A new paradigm for drug-induced torsadogenic risk assessment using human iPS cell-derived cardiomyocytes.
PMID 27956204
Abstract
Human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs) are anticipated to be a useful tool for conducting proarrhythmia risk assessments of drug candidates. However, a torsadogenic risk prediction paradigm using hiPSC-CMs has not yet been fully established. Extracellular field potentials (FPs) were recorded from hiPSC-CMs using the multi-electrode array (MEA) system. The effects on FPs were evaluated with 60 drugs, including 57 with various clinical torsadogenic risks. Actual drug concentrations in medium were measured using the equilibrium dialysis method with a Rapid Equilibrium Dialysis device. Relative torsade de pointes (TdP) scores were determined for each drug according to the degree of FP duration prolongation and early afterdepolarization occurrence. The margins were calculated from the free concentration in medium and free effective therapeutic plasma concentration. Each drug's results were plotted on a two-dimensional map of relative TdP risk scores versus margins. Each drug was categorised as high, intermediate, or low risk based on its location within predefined areas of the two-dimensional map. We categorised 19 drugs as high risk; 18 as intermediate risk; and 17 as low risk. We examined the concordance between our categorisation of high and low risk drugs against the torsadogenic risk categorisation in CredibleMeds®. Our system demonstrated high concordance, as reflected in a sensitivity of 81%, specificity of 87%, and accuracy of 83%. These results indicate that our torsadogenic risk assessment is reliable and has a potential to replace the hERG assay for torsadogenic risk prediction, however, this system needs to be improved for the accurate of prediction of clinical TdP risk. Here, we propose a novel drug induced torsadogenic risk categorising system using hiPSC-CMs and the MEA system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5060
E-4031, ≥98% (HPLC), solid
C21H27N3O3S · 2HCl
CDS023093
Nilotinib
C28H22F3N7O