EMAIL THIS PAGE TO A FRIEND
Nigerian journal of clinical practice
Can serums be replaced by Mueller-Hinton agar in germ tube test?
PMID 27958248
Abstract
The germ tube test (GTT) is inexpensive, easy, and well-defined test that differentiates Candida albicans (excluding Candida dubliniensis and Candida africana) from other species. The aim of this study was to evaluate various serums (i.e., human, rabbit, horse, and fetal bovine serum) used in the GTT and Mueller-Hinton agar (MHA). Fifty species isolated from various clinical samples that were defined as C. albicans by both conventional and DNA sequence analysis methods were included in the study. One to two colonies of C. albicans were mixed into 0.5-1 ml of fetal bovine serum, horse serum, rabbit serum, and human serum. Serums and MHA were incubated at 37°C for GTT. They were removed from the incubator and evaluated after 30 min, 1 h, 2 h, and 3 h of incubation. The GTT was accepted to be positive only if germ tube was 1/2 the width and 3 times the length of the parent yeast cell and with no constriction at the point of origin. When the use of serums and MHA for GTT was statistically evaluated, according to the positive scoring, the best results were obtained with MHA and with rabbit, horse, and fetal bovine serum, respectively. The best definition over time statistically was the third hour. It is suggested that inexpensive MHA is a fast, appropriate, and reliable medium for the probable diagnosis of GTT and C. albicans; however, additional studies are still needed to define other Candida species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
674974
6-Mercaptohexanoic acid, 90%
C6H12O2S