EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Matrine‑induced apoptosis in Hep3B cells via the inhibition of MDM2.
PMID 27959389
Abstract
Matrine, an alkaloid component derived from the Sophora root, can inhibit cancer cell proliferation and induce autophagy via p53xa0associated pathways. However, numerous tumor cells lack functional p53 and little is known about the effect of matrine on the p53‑deficient/mutant cancer cells. The present study aimed to assess anticancer effects of matrine in p53‑deficient human Hep3B hepatoma cells. The present results demonstrated that matrine caused Hep3B cell apoptosis by suppressing gene expression of minute double‑mutant (MDM)2. Notably, it was revealed that matrine inhibited MDM2xa0at the transcriptional level in a time‑ and dose‑dependent manner. This MDM2xa0inhibition resulted in induction of the p53xa0family member, p73; however, the functions of p73xa0were not induced since matrine‑induced p73xa0failed to activate its target genes, p21 and p53xa0upregulated modulator of apoptosis. The matrine‑induced downregulation of MDM2xa0led to an inhibition of inhibitor of apoptosis proteinxa03, which might serve a critical role in matrine‑induced apoptosis in MDM2‑overexpressing Hep3B cells. Finally, combination therapy of matrine withxa0100xa0µMxa0epotoside successfully killed more Hep3B cells, suggesting that matrine can sensitize p53‑deficient Hep3B cells to epotoside‑induced apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0752
Adenosine 5′-diphosphoribose sodium salt, ≥93%
C15H23N5O14P2