EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Engineering of the human vessel wall with hair follicle stem cells in vitro.
PMID 27959397
Abstract
Hair follicle stem cells (HFSCs) are increasingly used as a stem cell paradigm in vascular tissue engineering due to the fact that they are a rich source of easily accessible multipotent adult stem cells. Promising results have been demonstrated with small diameter (less than 6xa0mm) tissue engineered blood vessels under low blood pressure, however engineering large vessels (>6xa0mm in diameter) remains a challenge due to the fact it demands a higher number of seed cells and higher quality biomechanical properties. The aim of the current study was to engineer a large vessel (6xa0mm in diameter) with differentiated smooth muscle cells (SMCs) induced from human (h)HFSCs using transforming growth factor‑β1 and platelet‑derived growth factorxa0BB in combination with low‑serum culture medium. The cells were seeded onto polyglycolic acid and then wrapped around a silicone tube and further cultured inxa0vitro. A round vessel wall was formed subsequent to 8xa0weeks of culture. Histological examination indicated that layers of smooth muscle‑like cells and collagenous fibres were oriented in the induced group. In contrast, disorganised cells and collagenous fibres were apparent in the undifferentiated group. The approach developed in the current study demonstrated potential for constructing large muscular vessels with differentiated SMCs induced from hHFSCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAK008
Hydroxyproline Assay Kit, sufficient for 100 colorimetric tests