EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
The intrinsic stabilities and structures of alkali metal cationized guanine quadruplexes.
PMID 27966704
Abstract
The structures and stabilities of self-assembled guanine quadruplexes, M(9eG)8(+) (M = Na, K, Rb, Cs; 9eG = 9-ethylguanine), have been studied in the gas phase by blackbody infrared radiative dissociation to determine the difference in the stabilizing effect of the alkali metal cations. The order of stabilities to decomposition was determined to be K(+) > Rb(+) > Cs(+) ≫ Na(+), which is consistent with the observation of K(+) being the ion of choice in guanine quadruplexes in nucleic acids. In the gas phase, the sodiated quadruplex was found to lose one 9eG at a time, whereas the quadruplexes of the heavier cations lost a neutral guanine tetrad. Vibrational spectroscopy on the gas-phase quadruplex ions was consistent with the structures in which the metal cations were sandwiched between two guanine tetrads. Electronic structure calculations are also used to compare with the observed stabilities and vibrational spectra.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E4267
9-Ethylguanine
C7H9N5O