EMAIL THIS PAGE TO A FRIEND
Water science and technology : a journal of the International Association on Water Pollution Research
Post-treatment of palm oil mill effluent (POME) using combined persulphate with hydrogen peroxide (S2O8(2-)/H2O2) oxidation.
PMID 27973372
Abstract
The aim of the current study is to evaluate the effectiveness of combined persulphate with hydrogen peroxide (S2O8(2-)/H2O2) oxidation as a post-treatment of biologically treated palm oil mill effluent (POME) for the first time in the literature. The removal efficiencies of chemical oxygen demand (COD), ammoniacal nitrogen (NH3-N), and suspended solids (SS) were 36.8%, 47.6%, and 90.6%, respectively, by S2O8(2-) oxidation alone under certain operation conditions (i.e., S2O8(2-) = 0.82 g, pH 11, and contact time 20 min). Nevertheless, the combined process (S2O8(2-)/H2O2) achieved 75.8% and 87.1% removals of NH3-N and SS, respectively, under 2.45/1.63 g/g H2O2/S2O8(2-), pH 11, and 20 min oxidation. Moreover, 56.9% of COD was removed at pH 8.4.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
29580
cis,cis-1,5-Cyclooctadiene, ≥98.0% (GC)
C8H12