EMAIL THIS PAGE TO A FRIEND
The EMBO journal
SHARPIN regulates collagen architecture and ductal outgrowth in the developing mouse mammary gland.
PMID 27974362
Abstract
SHARPIN is a widely expressed multifunctional protein implicated in cancer, inflammation, linear ubiquitination and integrin activity inhibition; however, its contribution to epithelial homeostasis remains poorly understood. Here, we examined the role of SHARPIN in mammary gland development, a process strongly regulated by epithelial-stromal interactions. Mice lacking SHARPIN expression in all cells (Sharpin(cpdm)), and mice with a stromal (S100a4-Cre) deletion of Sharpin, have reduced mammary ductal outgrowth during puberty. In contrast, Sharpin(cpdm) mammary epithelial cells transplanted inxa0vivo into wild-type stroma, fully repopulate the mammary gland fat pad, undergo unperturbed ductal outgrowth and terminal differentiation. Thus, SHARPIN is required in mammary gland stroma during development. Accordingly, stroma adjacent to invading mammary ducts of Sharpin(cpdm) mice displayed reduced collagen arrangement and extracellular matrix (ECM) stiffness. Moreover, Sharpin(cpdm) mammary gland stromal fibroblasts demonstrated defects in collagen fibre assembly, collagen contraction and degradation inxa0vitro Together, these data imply that SHARPIN regulates the normal invasive mammary gland branching morphogenesis in an epithelial cell extrinsic manner by controlling the organisation of the stromal ECM.