EMAIL THIS PAGE TO A FRIEND
Materials science & engineering. C, Materials for biological applications
Enhancing osteogenic differentiation of MC3T3-E1 cells by immobilizing RGD onto liquid crystal substrate.
PMID 27987796
Abstract
To understand the effects of GRGDF modification on MC3T3-E1 cell behavior, we cultured these cells onto a biomimetic liquid crystalline matrix modified with GRGDF peptide (OPC-GA-RGD). Successful immobilization of GRGDF on the liquid crystalline surface was verified by fluorescent labeling, attenuated total reflectance Fourier transform infrared (ATR-FTIR) spectroscopy and X-ray photoelectron spectroscopy (XPS). OPC-GA-RGDs retained its liquid crystalline feature after surface modification. The RGD-immobilized OPC substrate was hardly beneficial to initial cell adhesion but could support long-term cell survival. The enhancement in cell proliferation did not correlate with RGD density. The lower GRGDF density immobilized on the liquid crystalline OPC matrix (OPC-GA-RGD3) promoted cell adhesion, proliferation, ALP expression level and mineralization, suggesting that both the viscoelasticity-based mechanical stimuli and receptor/ligand-based biochemical cue synergistically modulate MC3T3-E1 cell behavior.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O4733
Octanoyl chloride, 99%
C8H15ClO